Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Chlamydia trachomatis LGVII 434 External Run Controls
GSA_VASDP

Catalog No. 22157773 Shop All ZeptoMetrix Corporation Products
Click to view available options
Concentration:
5000 copies/mL
50000 copies/mL

Catalog No. 22-157-773

Supplier: ZeptoMetrix NATCT(434)ERCL

Only null left
Add to Cart
Add to Cart

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Contains 6 x 1.0mL vials of C. trachomatis NATtrol™ at concentrations of 5000 to 50,000 copies/mL
  • Supplied in purified protein matrix identical to the composition of true clinical specimen

Contain intact organisms and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO, 13485
Concentration 5000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol External Run Control
Formulation Formulated with purified, intact bacterial cells
For Use With (Application) Determination of presence of C. trachomatis DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.