Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ GBS External Run Controls
GSA_VASDP

Catalog No. p-4660123 Shop All ZeptoMetrix Corporation Products
Click to view available options
Format:
GBS Negative Result
GBS Positive Result

Catalog No. 21-100-548

Supplier: ZeptoMetrix NATLAC6MC

Only null left
Add to Cart
Add to Cart

For determination of the presence of Group B Streptococci DNA

  • Group B Streptococci (GBS) external run controls formulated with purified, intact and chemically modified organisms
  • Non infectious and refrigerator stable
  • Designed as full process extraction and amplification controls to validate molecular testing
  • Supplied in purified protein matrix that mimics composition of true clinical specimen
  • Evaluates performance of nucleic acid test
  • 6 x 0.5ml vials (each control)

For quality control of clinical assays and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Format GBS Negative Result
Media Type Vial
Product Type NATtrol External Run Control
Formulation Formulated with purified, intact bacterial cells (L.acidophilus Z048)
For Use With (Application) Determination of presence of group B Streptococci (GBS) DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.