Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Mycoplasma pneumoniae External Run Control
SDP

Catalog No. p-4597654 Shop All ZeptoMetrix Corporation Products
Click to view available options
Concentration:
50,000 copies/mL
5000 copies/mL

Formulated with intact chemically modified bacteria to render them non-infectious and refrigerator stable

  • Each run control pack contains 6 x 1mL vials of
      M. pneumoniae
    NATtrol™ at concentrations of 5,000 to 50,000 copies/mL
  • Supplied in purified serum protein matrix identical to the composition of true clinical specimen
  • Validated using an in-house real time PCR assay

Controls should be run using the same protocols as those used to run clinical specimens.

Used to validate extraction and amplification of M. pneumoniae in human serum or plasma

Specifications

Concentration 5000 copies/mL
Type NATtrol™ M. pneumoniae External Run Control
For Use With (Application) Used to validate extraction and amplification of M. pneumoniae in Human Serum or Plasma
Quantity 6 x 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.