Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Influenza A H1N1 External Run Controls

Click to view available options
Concentration:
25 to 28 Ct
28 to 31 Ct
Description
- Each control pack contains 6 x 1.0mL vials of Influenza A H1N1 NATtrol™ at concentrations 25 to 31 Ct Range
- Supplied in purified protein matrix identical to the composition of true clinical specimens
- Designed to evaluate the performance of nucleic acid tests for determination of the presence of Influenza A H1N1 RNA
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
Content And Storage | 2°C to 8°C |
Concentration | 28 to 31 Ct |
Detectable Analytes | Influenza A H1 RNA |
Type | Influenza A H1N1 External Run Control |
Quantity | 6 x 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction