Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Influenza External Run Controls
GSA_VASDP

Catalog No. 22156723 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Detectable Analytes:
Coxsackie virus A9 Nucleic Acid
Influenza A/NY/02/09 Nucleic Acid
Influenza A/NewCaledonia/20/99 Nucleic Acid, Influenza B/Florida/02/07 Nucleic Acid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-156-723 Coxsackie virus A9 Nucleic Acid
22-156-730 Influenza A/NewCaledonia/20/99 Nucleic Acid, Influenza B/Florida/02/07 Nucleic Acid
22-156-733 Influenza A/NY/02/09 Nucleic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-156-723 Supplier ZeptoMetrix Supplier No. NATCXVA96MC
Add to Cart
Add to Cart

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Each control contains 6 x 0.5mL vials of NATtrol™ Influenza A/B or Influenza A H1N1 (2009) or Coxsackie virus A9
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate the performance of nucleic acid tests for determination of presence of respiratory virus nucleic acids

Contain intact organisms and should be run in a manner identical to that used for clinical specimens.

Quality control of clinical assays and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Coxsackie virus A9 Nucleic Acid
Type Influenza External Run Control
For Use With (Application) Quality control of clinical assays and training of laboratory personnel
Quantity 6 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.