Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Purified Viral Lysate, Human Immunodeficiency Virus Type 1 (HIV-1)
SDP

Catalog No. 22157412
Click to view available options
Product Type:
Purified Viral Lysate

Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies

  • Available in 1mg vials

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration 0.5 to 3mg/mL
Content And Storage -65°C
Detects Viral Antibodies
Form Solution
Media Type Vial
Product Type Purified Viral Lysate
For Use With (Application) Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
Purification Method Sucrose density gradient ultracentrifugation
Quantity 1 mg
Strain Designations BaL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.