Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix RETROtek HIV-1 p24 Antigen ELISA
GSA_VASDP

Catalog No. 22156702
Click to view available options
Quantity:
5 x 96 Determinations
96 Determinations
Contains sodium azide as a preservative.
Contains sodium azide as a preservative.

Includes

HIV-1 p24 Antibody Coated Microplate for 96 determinations (1 plate), HIV-1 p24 Detector Antibody (1 vial) lyophilized, 25mL detector antibody diluent, 0.5 mL HIV-1 p24 Antigen Standard, 5mL lysing buffer, 0.3mL Streptavidin-Peroxidase, 100mL assay diluent, 125mL of 10X plate wash buffer, 0.3mL substrate,50mL substrate buffer, 12mL stop solution, 10 plate sealers, 1 resealable plastic bag.

Detects Human Immunodeficiency Virus Type 1 (HIV-1) p24 antigen in research specimens

  • Can be used to monitor the purification and biochemical behavior of HIV-1
  • May augment or supplant reverse transcriptase measurements traditionally employed to detect the presence of HIV-1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type RETROtek HIV-1 p24 Antigen ELISA
Sufficient For 5 x 96 Microplate Wells
For Use With (Application) Used to detect HIV-1 p24 Antigen in research specimens, including cell culture media, Human Sera and Plasma
Quantity 5 x 96 Determinations
Storage Requirements 2°C to 8°C

For research use only. Not for in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.