Learn More
ZeptoMetrix RETROtek SIV p27 Antigen ELISA Kit

Description
Includes
SIV p27 Antibody Coated Microplate for 96 determinations,1 plate: 12 x 8 well strips
SIV p27 Detector Antibody, 1 vial, lyophilized: Contains biotin-labelled polyclonal antibody to SIV, milk, Tween™, normal human serum and PBS
Detector Antibody Diluent: 25mL
SIV p27 Antigen Standard, 0.5mL: Contains detergent-disrupted, heat-inactivated viral antigen, goat serum, TritonX-100™ and sodium azide
Lysing Buffer, 5mL: Triton X-100™ in PBS and 2-chloroacetamide
Streptavidin peroxidase, 0.3mL: Contains streptavidin conjugated to horseradish peroxidase, PBS, goat serum, and 2-chloroacetamide
Assay Diluent, 100mL: Contains goat serum, PBS, Triton X-100™ and 2-chloroacetamide
10X Plate Wash Buffer, 125mL: Contains PBS, Tween 20™ and 2-chloroacetamide
Substrate, 0.3mL: Contains tetramethylbenzidine (TMB) and dimethyl sulfoxide
Substrate Buffer, 50mL: Contains citrate/acetate buffer, hydrogen peroxide and 2-chloroacetamide.
Requires
Disposable gloves, Validated adjustable micropipettes, single and multichannel, Test tubes and racks for preparing specimen and control dilutions, Graduated cylinders and assorted beakers, Validated automatic microplate washer or manual vacuum aspiration equipment, Validated incubator for 37°C ±1°C, Validated microplate reader, Timer, 1% sodium hypochlorite as disinfectant. May be prepared from household bleach, Distilled or deionized water
- Enzyme linked immunoassay used to monitor the purification and biochemical behavior of SIV
Specifications
Specifications
Product Type | RETROtek SIV p27 Antigen ELISA kit |
Sufficient For | 96-well Microplate |
For Use With (Application) | Used to detect Simian Immunodeficiency Virus (SIV) p27 Antigen in research specimens, including cell culture media, It can also be used to monito r the purification and biochemical behavior of SIV |
Kit Contents | Biotin-labelled Polyclonal Antibody to SIV, milk, Tween™, normal Human Serum, PBS, Detector antibody diluent, SIV p27 antibody coated microplate, 1 plate: 12 x 8-well strips, SIV p27 Detector Antibody, 1 vial, lyophilized |
Quantity | 96 Determinations |
Storage Requirements | 2°C to 8°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.