Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Clear PET-G Bottle
Natural Poly Bottle
Specifications
Specifications
Packaging | Clear PET-G Bottle |
CAS Min % | 93.0, 4.67, 0.0023 |
CAS Max % | 97.0, 4.86, 0.0023 |
Physical Form | Liquid |
Color | Colorless to Yellow |
Melting Point | Approximately 0°C |
Quantity | 500 mL |
Chemical Name or Material | Zero Oxygen Standard, For Calibration of Dissolved Oxygen Meters |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction