Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zeta Opioid Receptor Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297488APC
Description
Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Zeta Opioid Receptor | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor | |
Produced in rabbits immunized with E. coli-derived Human Zeta Opioid Receptor fragment. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
11054 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction