Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zeta Opioid Receptor Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299484MFV610
Description
Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Zeta Opioid Receptor | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Zeta Opioid Receptor. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
11054 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction