Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFAND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554782
Description
Immunogen sequence: QHCQVEHCRQ RDFLPFVCDD CSGIFCLEHR SRESHGCPEV TVINERLKTD QHTSYPCSFK DCAERELVAV ICP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 85%.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZFAND1 (AN1-type zinc finger protein 1) is a 268 amino acid protein that contains 2 AN1-type zinc fingers, which are often found in proteins that contain a ubiquitin-like domain and therefore play a role in the ubiquitination pathway. AN1-type zinc fingers contains six conserved cysteines and two histidines and have a dimetal (zinc)-bound alpha/beta fold. There are two isoforms of ZFAND1 that are produced as a result of alternative splicing events.
Specifications
ZFAND1 | |
Polyclonal | |
Unconjugated | |
ZFAND1 | |
AN1-type zinc finger protein 1; FLJ14007; ZFAN1; ZFAND1; zinc finger AN1-type containing 1; Zinc finger AN1-type-containing protein 1; zinc finger, AN1-type domain 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79752 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TCF1 | |
ZFAND1 | |
Recombinant protein corresponding to Human ZFAND1. Recombinant protein control fragment (Product #RP-93750). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction