Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFAND2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551274
Description
The antibody detects endogenous levels of total ZFAND2A protein.
ZFAND2A (AN1-type zinc finger protein 2A) is a 171 amino acid protein containing two AN1-type zinc fingers. AN1-type zinc fingers contain six conserved cysteines, two histidines and have a dimetal (zinc)-bound alpha/beta fold. The gene encoding ZFAND2A maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome.
Specifications
ZFAND2A | |
Polyclonal | |
Unconjugated | |
ZFAND2A | |
AIRAP; AN1-type zinc finger protein 2A; arsenite inducible RNA associated protein; ZFAND2A; zinc finger AN1-type containing 2A; zinc finger, AN1-type domain 2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90637 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
5.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N6M9 | |
ZFAND2A | |
Synthetic peptide of human ZFAND2A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction