Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFAND2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553995 Supplier Invitrogen™ Supplier No. PA553995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGQIPDVVVG DHIDRDCDSH PGKKKEKIFT YRCSKEGCKK KEMLQMVCAQ CHGNFCIQHR HPLDHSCRHG SRPTIKAG Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.

ZFAND2A (AN1-type zinc finger protein 2A) is a 171 amino acid protein containing two AN1-type zinc fingers. AN1-type zinc fingers contain six conserved cysteines, two histidines and have a dimetal (zinc)-bound alpha/beta fold. The gene encoding ZFAND2A maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFAND2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFAND2A
Gene Accession No. Q8N6M9
Gene Alias AIRAP; AN1-type zinc finger protein 2A; arsenite inducible RNA associated protein; ZFAND2A; zinc finger AN1-type containing 2A; zinc finger, AN1-type domain 2A
Gene Symbols ZFAND2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFAND2A. Recombinant protein control fragment (Product #RP-92808).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.