Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFAND2B Antibody (OTI1G10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274917C
Description
ZFAND2B Monoclonal specifically detects ZFAND2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
ZFAND2B | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZFAND2B | |
Full length recombinant protein of human ZFAND2B (NP_620157) produced in HEK293T cells. | |
0.1 mL | |
130617.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1G10 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
AIRAP-like protein, AN1-type 2B, AN1-type zinc finger protein 2B, arsenite inducible RNA associated protein-like, Arsenite-inducible RNA-associated protein-like protein, zinc finger, AN1-type domain 2B | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction