Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFAND2B Monoclonal Antibody (OTI2G10)
GREENER_CHOICE

Catalog No. PIMA525441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525441 Supplier Invitrogen™ Supplier No. MA525441
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZFAND2B Monoclonal Antibody for Western Blot, Flow

This gene encodes a protein containing AN1-type zinc-fingers and ubiquitin-interacting motifs. The encoded protein likely associates with the proteosome to stimulate the degradation of toxic or misfolded proteins. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFAND2B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2G10
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZFAND2B
Gene Accession No. Q8WV99
Gene Alias 1110060O18Rik; AIRAPL; AIRAP-like protein; AN1-type zinc finger protein 2B; arsenite-inducible RNA-associated protein-like protein; C81256; ZFAND2B; zinc finger AN1-type containing 2B; zinc finger, AN1 type domain 2B; zinc finger, AN1-type domain 2B
Gene Symbols ZFAND2B
Host Species Mouse
Immunogen Full length human recombinant protein of ZFAND2B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.