Learn More
Invitrogen™ ZFAND2B Monoclonal Antibody (OTI3A5)

Description
ZFAND2B Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | ZFAND2B |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI3A5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZFAND2B |
| Gene Accession No. | Q8WV99 |
| Gene Alias | 1110060O18Rik; AIRAPL; AIRAP-like protein; AN1-type zinc finger protein 2B; arsenite-inducible RNA-associated protein-like protein; C81256; ZFAND2B; zinc finger AN1-type containing 2B; zinc finger, AN1 type domain 2B; zinc finger, AN1-type domain 2B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.