Learn More
ZFAND2B Mouse anti-Human, Clone: OTI1G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502363

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein containing AN1-type zinc-fingers and ubiquitin-interacting motifs. The encoded protein likely associates with the proteosome to stimulate the degradation of toxic or misfolded proteins. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
ZFAND2B | |
Monoclonal | |
Unconjugated | |
ZFAND2B | |
AIRAPL | |
Mouse | |
Affinity Chromatography | |
RUO | |
130617 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WV99 | |
ZFAND2B | |
Full length human recombit protein of human ZFAND2B produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.