Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFAND2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557183 Supplier Invitrogen™ Supplier No. PA557183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPLCNVPVPV ARGEPPDRAV GEHIDRDCRS DPAQQKRKIF TNKCERAGCR QREMMKLTCE RCSRNFCIKH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes a protein containing AN1-type zinc-fingers and ubiquitin-interacting motifs. The encoded protein likely associates with the proteosome to stimulate the degradation of toxic or misfolded proteins. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFAND2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFAND2B
Gene Accession No. Q8WV99
Gene Alias 1110060O18Rik; AIRAPL; AIRAP-like protein; AN1-type zinc finger protein 2B; arsenite-inducible RNA-associated protein-like protein; C81256; ZFAND2B; zinc finger AN1-type containing 2B; zinc finger, AN1 type domain 2B; zinc finger, AN1-type domain 2B
Gene Symbols ZFAND2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFAND2B. Recombinant protein control fragment (Product #RP-96165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.