Learn More
Invitrogen™ ZFAND3 Monoclonal Antibody (OTI1H10)

Description
ZFAND3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ZFAND3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZFAND3 |
| Gene Accession No. | Q9H8U3 |
| Gene Alias | AN1-type zinc finger protein 3; AW539211; RNF1; TEG-27; testis expressed gene 27; testis expressed sequence 27; Testis-expressed protein 27; testis-expressed sequence 27; TEX27; ZFAND3; zinc finger AN1-type containing 3; zinc finger, AN1-type domain 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.