Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFAND3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596693 Supplier Invitrogen™ Supplier No. PA596693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGDAGSERSK APSLPPRCPC GFWGSSKTMN LCSKCFADFQ KKQPDDDSAP STSNSQSDLF SEETTSDNNN TSITTPTLSP SQQPLPTELN VTSPSKEECG PCTDTAHVSL ITPTKRSCGT DSQSENEASP VKRPRLLENT ERSEETSRSK QKSRRRCFQC QTKLELVQQE LGSCRCGYVF CMLHRLPEQH DCTFDHMGRG REEAIMKMVK LDRKVGRSCQ RIGEGCS; Positive Samples: A-549, BT-474, 293T.

ZFAND3 (Zinc Finger AN1-Type Containing 3) is a Protein Coding gene and also known as Testis expressed sequence 27.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFAND3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZFAND3
Gene Accession No. Q497H0, Q5U2M7
Gene Alias AN1-type zinc finger protein 3; AW539211; RNF1; TEG-27; testis expressed gene 27; testis expressed sequence 27; Testis-expressed protein 27; testis-expressed sequence 27; TEX27; ZFAND3; zinc finger AN1-type containing 3; zinc finger, AN1-type domain 3
Gene Symbols ZFAND3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human ZFAND3 (NP_0687621).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21769, 361816
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.