Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFAND5 Antibody (OTI5H8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274922FR
Description
ZFAND5 Monoclonal specifically detects ZFAND5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZFAND5 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ZFAND5 | |
Full length recombinant protein of human ZFAND5 (NP_005998) produced in E.coli. | |
0.1 mL | |
7763.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5H8 | |
Western Blot, Immunohistochemistry | |
Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction