Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFAND5 Monoclonal Antibody (OTI5B8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800483

Description
ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.Specifications
ZFAND5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O76080 | |
ZFAND5 | |
Full length human recombit protein of human ZFAND5 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI5B8 | |
Unconjugated | |
ZFAND5 | |
ZA20D2, ZFAND5A, ZNF216 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7763 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction