Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFAND5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA553735
Description
Immunogen sequence: TASGSNSPTS DSASVQRADT SLNNCEGAAG STSEKSRNVP VAALPVTQQM TEMSISREDK ITTPKTEVSE PVVTQPSPSV SQPSTSQSEE KAPELPKPKK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.
Specifications
| ZFAND5 | |
| Polyclonal | |
| Unconjugated | |
| ZFAND5 | |
| 2310057A04Rik; 5830475F03Rik; AA415485; AN1-type zinc finger protein 5; EST00848 homolog; ubiquitin homolog; Za20d2; Zfand5; ZFAND5A; Zfp216; Zinc finger A20 domain-containing protein 2; zinc finger AN1-type containing 5; zinc finger protein 216; zinc finger, A20 domain containing 2; zinc finger, AN1-type domain 5; ZNF216 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76080 | |
| ZFAND5 | |
| Recombinant protein corresponding to Human ZFAND5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction