Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980713
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0713 Supplier Bioss Supplier No. BS10142R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.

Specifications

Antigen ZFAND5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZFAND5
Gene Accession No. O76080
Gene Alias 2310057A04Rik; 5830475F03Rik; AA415485; AN1-type zinc finger protein 5; BOS_8857; EST00848 homolog; LOC100344281; ZA20D2; Zfand5; ZFAND5A; Zfp216; Zinc finger A20 domain-containing protein 2; zinc finger AN1-type containing 5; Zinc finger protein 216; zinc finger, A20 domain containing 2; zinc finger, AN1-type domain 5; Znf216
Gene Symbols ZFAND5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZFAND5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7763
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.