Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFAND5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317328100UL
This item is not returnable.
View return policy
Description
ZFAND5 Polyclonal antibody specifically detects ZFAND5 in Human samples. It is validated for ImmunofluorescenceSpecifications
ZFAND5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: CSTGCGFYGNPRTNGMCSVCYKEHLQR | |
100 μg | |
Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
7763 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction