Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFC3H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552391 Supplier Invitrogen™ Supplier No. PA552391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSRKQNYSSK NENCVEETFE DLLLKYKQIQ LELECINKDE KLALSSKEEN VQEDPKTLNF EDQTSTDNVS ITKDSSKEVA PEEKTQVKTF QAFELKPLRQ KLTLPGDKNR LKKVKDGAKP LSL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 84%.

Subunit of the trimeric poly(A) tail exosome targeting (PAXT) complex, a complex that directs a subset of long and polyadenylated poly(A) RNAs for exosomal degradation. The RNA exosome is fundamental for the degradation of RNA in eukaryotic nuclei. Substrate targeting is facilitated by its cofactor MTREX, which links to RNA-binding protein adapters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFC3H1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFC3H1
Gene Accession No. O60293
Gene Alias BC033596; Ccdc131; coiled-coil domain containing 131; coiled-coil domain-containing protein 131; CSRC2; KIAA0546; MGC23401; proline/serine-rich coiled-coil 2; proline/serine-rich coiled-coil protein 2; Psrc2; ZFC3H1; Zinc finger C3H1 domain-containing protein; zinc finger C3H1-type containing; zinc finger, C3H1-type containing
Gene Symbols ZFC3H1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFC3H1. Recombinant protein control fragment (Product #RP-88604).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.