Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFC3H1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568154
Description
Predicted to react with Mouse samples.
Subunit of the trimeric poly(A) tail exosome targeting (PAXT) complex, a complex that directs a subset of long and polyadenylated poly(A) RNAs for exosomal degradation. The RNA exosome is fundamental for the degradation of RNA in eukaryotic nuclei. Substrate targeting is facilitated by its cofactor MTREX, which links to RNA-binding protein adapters.
Specifications
ZFC3H1 | |
Polyclonal | |
Unconjugated | |
ZFC3H1 | |
BC033596; Ccdc131; coiled-coil domain containing 131; coiled-coil domain-containing protein 131; CSRC2; KIAA0546; MGC23401; proline/serine-rich coiled-coil 2; proline/serine-rich coiled-coil protein 2; Psrc2; ZFC3H1; Zinc finger C3H1 domain-containing protein; zinc finger C3H1-type containing; zinc finger, C3H1-type containing | |
Rabbit | |
affinity chromatography | |
RUO | |
196441, 216345 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60293 | |
ZFC3H1 | |
Synthesized peptide derived from human ZFC3H1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction