Learn More
Invitrogen™ ZFC3H1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total CCDC131.
Specifications
Specifications
| Antigen | ZFC3H1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZFC3H1 |
| Gene Accession No. | O60293 |
| Gene Alias | BC033596; Ccdc131; coiled-coil domain containing 131; coiled-coil domain-containing protein 131; CSRC2; KIAA0546; MGC23401; proline/serine-rich coiled-coil 2; proline/serine-rich coiled-coil protein 2; Psrc2; ZFC3H1; Zinc finger C3H1 domain-containing protein; zinc finger C3H1-type containing; zinc finger, C3H1-type containing |
| Gene Symbols | ZFC3H1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.