Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFHX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568554 Supplier Invitrogen™ Supplier No. PA568554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 87%; Rabbit: 93%; Rat: 86%.

ZFHX2 gene ontology annotations related to this gene include adult behavior; regulation of sensory perception of pain; regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFHX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFHX2
Gene Accession No. Q9C0A1
Gene Alias Kiaa1056; KIAA1762; LOW QUALITY PROTEIN: zinc finger homeobox protein 2; retina zinc finger homeodomain protein; ZFH; ZFH-2; ZFH-5; ZFHX2; zinc finger homeobox 2; zinc finger homeobox protein 2; Zinc finger homeodomain protein 2; Zinc finger homeodomain protein 5; zinc finger protein 409; ZNF409
Gene Symbols ZFHX2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZFHX2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85446
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.