Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFHX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563707 Supplier Invitrogen™ Supplier No. PA563707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAQVQVPQQS HQQILPQQQQ NQLSIAQSHS ALLQPSQHPE KKNKLVIKEK EKESQRERDS AEGGEGNTGP KETLPDA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 76%.

Transcriptional repressor. It inhibits the enhancer element of the AFP gene by binding to its AT-rich core sequence. Regulator of myoblasts differentiation through the binding to the AT-rich sequence of MYF6 promoter and promoter repression. Down-regulates the MUC5AC promoter in gastric cancer. Miura Y., J. Biol. Chem. 270:26840-26848(1995). Morinaga T., Mol. Cell. Biol. 11:6041-6049(1991). Martin J., Nature 432:988-994(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFHX3
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFHX3
Gene Accession No. Q15911
Gene Alias A230102L03Rik; alpha-fetoprotein enhancer binding protein; alpha-fetoprotein enhancer-binding protein; AT motif binding factor 1; AT motif-binding factor 1; ATBF1; AT-binding transcription factor 1; ATBT; C16orf47; mKIAA4228; Putative uncharacterized protein C16orf47; RGD1560268; WBP9; ZFH-3; Zfhx3; zinc finger homeobox 3; zinc finger homeobox protein 3; Zinc finger homeodomain protein 3; ZNF927
Gene Symbols ZFHX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFHX3. Recombinant protein control fragment (Product #RP-101926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.