Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFHX4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$646.00
Specifications
Antigen | ZFHX4 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZFHX4 Polyclonal specifically detects ZFHX4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZFHX4 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ16514, FLJ20980, ZFH-4, ZFH4ZHF4, zinc finger homeobox 4, zinc finger homeobox protein 4, zinc finger homeodomain 4, Zinc finger homeodomain protein 4, zinc-finger homeodomain protein 4 | |
ZFHX4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q86UP3 | |
79776 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ESKDQENNCERPKESNVLHPNGECPVKSEPTEPGDEDEEDAYSNELDDEEVLGELTDSIGNKDFPLLNQSISPLSSSVLKFIEKGTSSSSATVSDDTEKKKQTAAVRASGSVASNYGISGKDFADA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title