Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFHX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599757 Supplier Invitrogen™ Supplier No. PA599757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZFHX4.

May play a role in neural and muscle differentiation. May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFHX4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZFHX4
Gene Accession No. Q86UP3, Q9JJN2
Gene Alias A930021B15; C130041O22Rik; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; RGD1563022; ZFH4; Zfh-4; Zfhx4; ZHF4; zinc finger homeobox 4; zinc finger homeobox protein 4; zinc finger homeobox protein 4; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; zinc finger homeodomain 4; zinc finger homeodomain protein 4; zinc-finger homeodomain protein 4
Gene Symbols ZFHX4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZFHX4(Accession Q86UP3), corresponding to amino acid residues D208-N258.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310250, 79776, 80892
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.