Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540404
Description
Peptide sequence: SNRSYAGKQT DECNEFGKAL LYLKQEKTHS GVEYSEYNKS GKALSHKAAI Sequence homology: Cow: 92%; Dog: 86%; Human: 100%; Mouse: 79%; Rat: 86%.
Zinc finger proteins (Zfp) are encoded by a large family of genes present in many organisms including yeast and human. Some of them are transcriptional activators and bind specifically to DNA by zinc mediated folded structures commonly known as zinc fingers. The putative Zfp-1 (Zinc finger protein 1 homolog) protein contains in addition to 7 zinc fingers, two helix-turn-helix motifs.
Specifications
| ZFP1 | |
| Polyclonal | |
| Unconjugated | |
| ZFP1 | |
| FLJ34243; Fnp-1; mkr-1; Protein mKR1; RGD1565049; ZFP1; Zfp-1; ZFP1 zinc finger protein; zinc finger protein 1; Zinc finger protein 1 homolog; zinc finger protein 475; ZNF475 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 162239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P2D0 | |
| ZFP1 | |
| Synthetic peptide directed towards the N-terminal of human ZFP1 (aa 122-171). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction