Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564077 Supplier Invitrogen™ Supplier No. PA564077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRSYAGKQTD ECNEFGKALL YLKQEKTHSG VEYSEYNKSG KALSHKAAIF KHQKIKNLVQ PFICTYCDK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 64%.

Zinc finger proteins (Zfp) are encoded by a large family of genes present in many organisms including yeast and human. Some of them are transcriptional activators and bind specifically to DNA by zinc mediated folded structures commonly known as zinc fingers. The putative Zfp-1 (Zinc finger protein 1 homolog) protein contains in addition to 7 zinc fingers, two helix-turn-helix motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFP1
Gene Accession No. Q6P2D0
Gene Alias FLJ34243; Fnp-1; mkr-1; Protein mKR1; RGD1565049; ZFP1; Zfp-1; ZFP1 zinc finger protein; zinc finger protein 1; Zinc finger protein 1 homolog; zinc finger protein 475; ZNF475
Gene Symbols ZFP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFP1. Recombinant protein control fragment (Product #RP-103478).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162239
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.