Learn More
ZFP106 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301526AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZFP106 (Zinc finger protein 106), also known as zinc finger protein 474, is a 1883 amino acid human homolog of the mouse Zfp106 protein and is a member of the Kruppel C2H2-type zinc-finger family. Localized to the nucleus, ZFP106 contains two C2H2-type zinc fingers and is thought to be involved in transcriptional regulation.Specifications
| ZFP106 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O88466, Q9H2Y7 | |
| Zfp106, ZNF106 | |
| Between 1300 and 1350 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ZNF106 | |
| SH3BP3, ZFP106, ZNF474 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20402, 64397 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.