Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551175
Description
The antibody detects endogenous levels of total ZFP2 protein.
Probable transcription factor involved in neuronal differentiation and/or phenotypic maintenance.
Specifications
ZFP2 | |
Polyclonal | |
Unconjugated | |
ZFP2 | |
9930007F06Rik; Fnp-2; Mkr2; mkr-2; mszf87; Protein mKR2; Zfp2; zfp-2; ZFP2 zinc finger protein; zinc finger protein 2; Zinc finger protein 2 homolog; zinc finger protein 751; ZNF751 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80108 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6ZN57 | |
ZFP2 | |
Fusion protein of human ZFP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction