Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP200 Antibody (OTI5F4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274923IR
Description
ZFP200 Monoclonal specifically detects ZFP200 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZFP200 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF200 | |
Full length recombinant protein of human ZFP200 (NP_932353) produced in E.coli. | |
0.1 mL | |
7752.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5F4 | |
Western Blot, Immunohistochemistry | |
MGC45293, zinc finger protein 200, ZNFMF | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction