Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569014 Supplier Invitrogen™ Supplier No. PA569014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%.

ZFP28 may be involved in transcriptional regulation. may have a role in embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP28
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFP28
Gene Accession No. Q8NHY6
Gene Alias 2810438M17Rik; KIAA1431; Krueppel-like zinc finger factor X6; LOW QUALITY PROTEIN: zinc finger protein 28 homolog; mkr5; mkr-5; Protein mKR5; ZFP28; zfp-28; ZFP28 zinc finger protein; Zfp28l; zinc finger factor X6; zinc finger protein 28; Zinc finger protein 28 homolog
Gene Symbols ZFP28
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZFP28.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140612
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.