Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556000
Description
Immunogen sequence: LLEQEKEPWM VKRELTGSLF SGQRSVHETQ ELFPKQDSYA EGVTDRTSNT KLDCSSFREN WDSDYVFGRK LAVGQETQFR QEPITHNKTL SKERERTYNK SGRWFYLDDS EEKVHNRDSI KNFQKSSVVI KQTGIYAG Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 46%.
ZFP28 may be involved in transcriptional regulation. may have a role in embryonic development.
Specifications
| ZFP28 | |
| Polyclonal | |
| Unconjugated | |
| ZFP28 | |
| 2810438M17Rik; KIAA1431; Krueppel-like zinc finger factor X6; LOW QUALITY PROTEIN: zinc finger protein 28 homolog; mkr5; mkr-5; Protein mKR5; ZFP28; zfp-28; ZFP28 zinc finger protein; Zfp28l; zinc finger factor X6; zinc finger protein 28; Zinc finger protein 28 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NHY6 | |
| ZFP28 | |
| Recombinant protein corresponding to Human ZFP28. Recombinant protein control fragment (Product #RP-99750). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction