Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Zfp352 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119722
This item is not returnable.
View return policy
Description
Positive Control: Non-transfected 293T cells lysate, GFP-tagged Zfp352 transfected 293T cells lysate, Non-transfected 293F cells lysate, GFP-tagged Zfp352 transfected 293F cells lysate. Subcellular Location: Nucleus.
Zfp352 (formerly 2czf48), that is expressed in early mouse embryos. The Zfp352 gene is intronless in the coding region but carries a solitary 4.3kb intron in the 5 untranslated region.
Specifications
| Zfp352 | |
| Polyclonal | |
| Unconjugated | |
| Zfp352 | |
| Zinc finger protein 352 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 236537 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| Zfp352 | |
| Synthetic peptide within mouse Zfp352 aa 100-150. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction