Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541450 Supplier Thermo Scientific Supplier No. PA541450
Add to Cart
Add to Cart

ZFP36 Polyclonal Antibody

ZFP36 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP36
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zfp36
Gene Accession No. P26651
Gene Alias G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog
Gene Symbols Zfp36
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZFP36.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7538
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.