Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP36L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209699
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9699 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9699 Supplier Fabgennix Inc Supplier No. ZFP36L1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFP36L1 Polyclonal antibody specifically detects ZFP36L1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ZFP36L1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ZFP36L1
Gene Accession No. P17431, P23950, Q07352
Gene Alias AW742437; AW743212; Berg36; Brf1; butyrate response factor 1; cMG1; D530020L18Rik; early response factor Berg36; EGF-inducible protein CMG1; EGF-response factor 1; ERF1; ERF-1; mRNA decay activator protein ZFP36L1; RNF162B; TIS11b; TPA-induced sequence 11b; ZFP36 ring finger protein like 1; ZFP36 ring finger protein-like 1; ZFP36L1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid reigon 200-250 on human Zinc finger protein 36, C3H1 type-like 1.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12192, 29344, 677
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.