Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP37 Monoclonal Antibody (OTI3E7), TrueMAB™, OriGene
SDP

Catalog No. 501683259
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3259 Supplier OriGene Supplier No. TA810209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZFP37 may be involved in transcriptional regulation.

Specifications

Antigen ZFP37
Applications Western Blot
Classification Monoclonal
Clone OTI3E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZFP37
Gene Accession No. Q9Y6Q3
Gene Alias ZNF906, zfp-37
Gene Symbols ZFP37
Host Species Mouse
Immunogen Full length human recombit protein of human ZFP37 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7539
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.