Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568601 Supplier Thermo Scientific Supplier No. PA568601
Add to Cart
Add to Cart

ZFP37 Polyclonal Antibody

ZFP37 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP37
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFP37
Gene Accession No. Q9Y6Q3
Gene Alias LOW QUALITY PROTEIN: zinc finger protein 37 homolog; male germ cell-specific zinc finger protein; Tzn; ZFP37; Zfp-37; ZFP37 zinc finger protein; zinc finger protein 37; Zinc finger protein 37 homolog; zinc finger protein homologous to Zfp37 in mouse; ZNF906
Gene Symbols ZFP37
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZFP37.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7539
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.