Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Zfp444 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568866
Description
This target displays homology in the following species: Human: 92%; Mouse: 100%; Rat: 100%.
Zfp444 gene ontology annotations related to this gene include positive regulation of transcription, DNA-templated.
Specifications
Zfp444 | |
Polyclonal | |
Unconjugated | |
Zfp444 | |
2810031J10Rik; 6230401O10Rik; mFLJ00134; Zfp444; zinc finger protein 444; Znf444 | |
Rabbit | |
Affinity chromatography | |
RUO | |
72667 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
Zfp444 | |
synthetic peptide corresponding to a region of mouse Zfp444. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction