Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP57 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299543PE
Description
ZFP57 Polyclonal antibody specifically detects ZFP57 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ZFP57 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
C6orf40, TNDM1, zfp-57, zinc finger protein 57 homolog (mouse), zinc finger protein 698, ZNF698 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ZFP57 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
346171 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction