Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP57 Monoclonal Antibody (GT865)
GREENER_CHOICE

Catalog No. PIMA527869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527869 Supplier Invitrogen™ Supplier No. MA527869
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 3xFlag-human ZFP57-transfected 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a zinc finger protein containing a KRAB domain. Studies in mouse suggest that this protein may function as a transcriptional repressor. Mutations in this gene have been associated with transient neonatal diabetes mellitus type 1 (TNDM1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP57
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT865
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ZFP57
Gene Accession No. Q9NU63
Gene Alias bA145L22; bA145L22.2; C6orf40; DADB-25P22.4; G19; TNDM1; ZFP57; Zfp-57; ZFP57 zinc finger protein; zinc finger protein 57; Zinc finger protein 57 homolog; zinc finger protein 698; ZNF698
Gene Symbols ZFP57
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ZFP57. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.