Learn More
Invitrogen™ Zfp57 Monoclonal Antibody (OTI1E2)

Description
Zfp57 Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | Zfp57 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ZFP57 |
| Gene Accession No. | Q8C6P8, Q9NU63 |
| Gene Alias | bA145L22; bA145L22.2; C6orf40; DADB-25P22.4; G19; TNDM1; ZFP57; Zfp-57; ZFP57 zinc finger protein; zinc finger protein 57; Zinc finger protein 57 homolog; zinc finger protein 698; ZNF698 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.