Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP64 Antibody (PCRP-ZFP64-1H2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308648MFV610
Description
ZFP64 Monoclonal antibody specifically detects ZFP64 in Human samples. It is validated for Flow CytometrySpecifications
ZFP64 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
55734 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-ZFP64-1H2 | |
Flow Cytometry | |
dJ548G19.1, dJ831D17.1, FLJ10734, FLJ12628, Zfp-64, Zinc finger protein 338MGC940, zinc finger protein 64, zinc finger protein 64 homolog (mouse), ZNF338 | |
Recombinant full-length human ZFP64 protein (Uniprot: Q9NTW7) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction